Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%

Supplier: Origen Biomedical, Inc

H62812-03 H62812-MD
AAH62812-03EA 380 USD InStock
AAH62812-03 AAH62812-MD
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester 96%
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%

Purity: 96% Molecular Formula: C13H18BFO3 Formula Weight: 252.09 Physical Form: Solid


Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996294
CAS Number: 1599432-41-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty