Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-(1-aminoethyl)phenol 97%

Supplier: Origen Biomedical, Inc
Danger

H63001-03 H63001-06 H63001-MD
AAH63001-03EA 201 USD InStock
AAH63001-03 AAH63001-06 AAH63001-MD
(R)-4-(1-aminoethyl)phenol 97%
(R)-4-(1-aminoethyl)phenol
(R)-4-(1-Aminoethyl)phenol, 97%

Purity: 97% CAS No.: 134855-89-3 Molecular Formula: C8H11NO Formula Weight: 137.18 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: White UN No.: 3259 MDL No.: MFCD03844647


Formula: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD03844647
CAS Number: 134855-89-3
UN: 3259
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty