Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-bromo-6-methylpyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H64301-03 H64301-06
AAH64301-03EA 69.6 USD InStock
AAH64301-03 AAH64301-06 AAH64301-MD
3-Amino-2-bromo-6-methylpyridine 98%
3-Amino-2-bromo-6-methylpyridine
3-Amino-2-bromo-6-methylpyridine, 98%

Purity: 98% CAS No.: 126325-53-9 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Off-white Beilstein Reference: 4243986 UN No.: 2811 MDL No.: MFCD03095078


Formula: C₆H₇BrN₂
MW: 187.04
Storage Temperature: Ambient
MDL Number: MFCD03095078
CAS Number: 126325-53-9
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty