Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-2-bromo-3-methylpyridine 98%

Supplier: Origen Biomedical, Inc
Warning

H66134-06 H66134-14
AAH66134-06EA 64.4 USD InStock
AAH66134-06 AAH66134-14
5-Amino-2-bromo-3-methylpyridine 98%
5-Amino-2-bromo-3-methylpyridine
5-Amino-2-bromo-3-methylpyridine, 98%

Purity: 98% CAS No.: 38186-83-3 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Grey Melting Point: 97-100° Beilstein Reference: 471832 UN No.: 2811 MDL No.: MFCD03095085


Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD03095085
CAS Number: 38186-83-3
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty