H66497-03
H66497-06
AAH66497-03EA
46.4
USD
InStock
AAH66497-03
AAH66497-06
1-tert-Butylpiperazine 97%
1-tert-Butylpiperazine
1-tert-Butylpiperazine, 97%
Purity: 97%
CAS No.: 38216-72-7
Molecular Formula: C8H18N2
Formula Weight: 142.24
Physical Form: Low melting solid
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Colorless to yellow
Melting Point: 32-35°
Boiling Point: 85-87°/22mm
UN No.: 3259
MDL No.: MFCD00049359