Merit™ Lister Bandage Scissors, Physician Grade, Sklar
Supplier: Sklar
97-222 97-237 97-226 97-231 97-241
10184-800EA 13.14 USD InStock
10184-800 10184-802 10192-456 10193-646 10193-656
Merit™ Lister Bandage Scissors, Physician Grade, Sklar
Scissors
Merit™ Lister bandage scissors are used for sizing dressings and removing circumferential bandages.
  • Angled jaws
  • Lower blade is slightly longer
  • Flattened blunt nodule on tip of lower blade
The jaws of the lister bandage scissors are angled with the lower blade being slightly longer
Merit™ instruments are office-grade line of products used for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a ten year guarantee.
The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows lister bandage scissors to be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty