Examination Gloves, Nitrile, Extra-Thick, Tronex®
Supplier: Tronex
9662-10 9662-20 9662-30 9662-35
10153-100BX 24.1 USD InStock
10153-100 10153-102 10153-104 10153-106
Examination Gloves, Nitrile, Extra-Thick, Tronex®
Gloves
Heavy-duty, extra-thick style with extended cuff and textured surface offers optimal strength, puncture resistance, and resistance to chemotherapy drugs.
  • Fully textured for sustained grip during use
  • Maximum durability and performance
  • Barrier protection from bloodborne pathogens
  • Thickness: 9.6 mil (finger)
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty