Rat Recombinant VEGF (from E. coli)
Supplier: PEPROTECH INC.
400-31-2UG 400-31-10UG 400-31-100UG 400-31-250UG 400-31-500UG 400-31-1MG
10771-046EA 132.81 USD InStock
10771-046 10771-048 10771-050 10771-052 10771-054 10771-056
Rat Recombinant VEGF (from E. coli)
Proteins and Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothellial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant Rat VEGF
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty