Human Recombinant VEGF (from E. coli)
Supplier: PEPROTECH INC.
100-20A-2UG 100-20A-10UG 100-20A-50UG 100-20A-100UG 100-20A-250UG 100-20A-500UG 100-20A-1MG
10771-984EA 143.88 USD InStock
10771-984 10771-986 10771-988 10771-990 10771-992 10771-994 10771-996
Human Recombinant VEGF (from E. coli)
Proteins and Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. VEGF signals through three receptors; fms- like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Due to its increased acidity, VEGF Recombinant Human VEGF
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty