Human Recombinant NOV (from E. coli)
Supplier: PEPROTECH INC.
120-26-5UG 120-26-20UG 120-26-100UG 120-26-250UG 120-26-500UG 120-26-1MG
10772-636EA 139.89 USD InStock
10772-636 10772-638 10772-640 10772-642 10772-644 10772-646
Human Recombinant NOV (from E. coli)
Proteins and Peptides
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm’s tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorgenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities. Recombinant Human NOV is a 36.2 kDa protein containing 331 amino acid residues. It is composed of four distinct structural domains (modules): the IGF binding protein (IGFBP) domain; the von Willebrand Factor C (VWFC) domain; the Thrombospondin type-I (TSP type-1) domain; and a C-terminal cysteine knot-like domain (CTCK).
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty