Human Recombinant IL-8 (from E. coli)
Supplier: PEPROTECH INC.
200-08M-5UG 200-08M-25UG 200-08M-50UG 200-08M-100UG 200-08M-250UG 200-08M-500UG 200-08M-1MG
10772-956EA 139.89 USD InStock
10772-956 10772-958 10772-960 10772-962 10772-964 10772-966 10772-968
Human Recombinant IL-8 (from E. coli)
Proteins and Peptides
IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant Human IL-8 (CXCL8) (monocyte-derived) is an 8.4 kDa protein containing 72 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty