Mouse Recombinant Exodus-2 (from E. coli)
Supplier: PEPROTECH INC.
250-13-5UG 250-13-20UG 250-13-100UG 250-13-250UG 250-13-500UG 250-13-1MG
10773-204EA 139.89 USD InStock
10773-204 10773-206 10773-208 10773-210 10773-212 10773-214
Mouse Recombinant Exodus-2 (from E. coli)
Proteins and Peptides
Exodus-2 is a CC chemokine that can signal through the CCR7 receptor. It is expressed in lymph nodes of certain endothelial cells, and in the spleen and appendix. Exodus-2 chemoattracts T and B lymphocytes and inhibits hematopoiesis. Exodus-2 contains six cysteine residues, including the four conserved cysteines present in CC chemokines. Recombinant Murine Exodus-2 (CCL21) is a 12.0 kDa protein containing 110 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty