Human Recombinant IL-8 (from E. coli)
Supplier: PEPROTECH INC.
AF-200-08M-5UG AF-200-08M-25UG AF-200-08M-100UG AF-200-08M-250UG
10778-306EA 183.06 USD InStock
10778-306 10778-308 10778-310 10778-312
Human Recombinant IL-8 (from E. coli)
Proteins and Peptides
IL-8 is a proinflammatory CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is secreted by monocytes and endothelial cells. IL-8 chemoattracts and activates neutrophils. Recombinant Human IL-8 (CXCL8) (monocyte-derived) is an 8.4 kDa protein containing 72 amino acid residues.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty