Human Recombinant IL-17A (from E. coli)
Supplier: PEPROTECH INC.
AF-200-17-5UG AF-200-17-25UG AF-200-17-100UG AF-200-17-250UG AF-200-17-500UG AF-200-17-1MG
10778-358EA 183.06 USD InStock
10778-358 10778-360 10778-362 10778-364 10778-366 10778-368
Human Recombinant IL-17A (from E. coli)
Proteins and Peptides
The originally described IL-17 protein, now known as IL-17A, is a homodimer of two 136 amino acid chains that are secreted by activated T-cells, which act on stromal cells to induce production of proinflammatory and hematopoietic bioactive molecules. Today, IL-17 represents a family of structurally-related cytokines that share a highly conserved C-terminal region, but differ from one another in their N-terminal regions and in their distinct biological roles. The six known members of this family, IL-17A through IL-17F, are secreted as homodimers. IL-17A exhibits cross-species bioactivity between human and murine cells. Recombinant Human IL-17A is a 31.3 kDa, disulfide-linked homodimer of two 137 amino acid, polypeptide chains.
  • Animal-free
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty