Human Recombinant FGF-acidic (from E. coli)
Supplier: PEPROTECH INC.
100-17A-10UG 100-17A-50UG 100-17A-100UG 100-17A-250UG 100-17A-500UG 100-17A-1MG
10778-886EA 145.16 USD InStock
10778-886 10778-888 10778-890 10778-892 10778-894 10778-896
Human Recombinant FGF-acidic (from E. coli)
Proteins and Peptides
FGF-acidic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-acidic is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. FGF-acidic has the ability to signal through all the FGF receptors. Recombinant Human FGF-acidic is a 16.8 kDa protein consisting of 141 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty