Human Recombinant EG-VEGF (from E. coli)
Supplier: PEPROTECH INC.
100-44-5UG 100-44-20UG 100-44-100UG 100-44-250UG 100-44-500UG 100-44-1MG
10779-002EA 143.88 USD InStock
10779-002 10779-004 10779-006 10779-008 10779-010 10779-012
Human Recombinant EG-VEGF (from E. coli)
Proteins and Peptides
EG-VEGF is a secreted angiogenetic mitogen growth factor expressed in the steroidogenic glands, ovary, testis, adrenal gland, and placenta.
  • Recombinant Human EG-VEGF is a 9.6 kDa protein consisting of 86 amino acid residues, including ten cysteine residues that potentially form five pairs of intra-molecular disulfide bonds
EG-VEGF induces proliferation, migration, and fenestration (formation of membrane discontinuities) in capillary endothelial cells derived from endocrine glands. The human EG-VEGF gene codes for a 105 amino acid polypeptide containing an N-terminal signal sequence of 19 amino acids.
Precaución: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty