Human Recombinant Fetuin (from HEK293 cells)
Supplier: PEPROTECH INC.
140-13-10UG 140-13-50UG 140-13-100UG 140-13-250UG 140-13-500UG 140-13-1MG
10779-444EA 132.81 USD InStock
10779-444 10779-446 10779-448 10779-450 10779-452 10779-454
Human Recombinant Fetuin (from HEK293 cells)
Proteins and Peptides
Fetuin A/AHSG is a human plasma glycoprotein belonging to the Cystatin family of protease inhibitors. It is highly expressed in embryonic cells and adult hepatocytes, and is expressed to a lesser extent in monocytes/macrophages. Fetuin A/AHSG is a major serum protein component that exerts various calcium-dependent physiological activities, and can mediate growth signaling in certain tumor cells. It also can act as a natural antagonist against specific TGF-β and BMP signaling proteins. Recombinant Human Fetuin A is a heterodimeric protein containing a 282 amino acid A chain and a 27 amino acid B chain. The calculated molecular weight of Recombinant Human Fetuin A/AHSG is 32.9 kDa. Due to glycosylation, Recombinant Human Fetuin A/AHSG migrates at an apparent molecular weight of 45–55 kDa by SDS-PAGE run under non-reducing conditions.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty