Human Recombinant PECAM-1 (from HEK293 Cells)
Supplier: PEPROTECH INC.
150-06-10UG 150-06-50UG 150-06-100UG 150-06-250UG 150-06-500UG 150-06-1MG
10779-500EA 132.81 USD InStock
10779-500 10779-502 10779-504 10779-506 10779-508 10779-510
Human Recombinant PECAM-1 (from HEK293 Cells)
Proteins and Peptides
PECAM is transmembrane glycoprotein that belongs to the Ig-related superfamily of adhesion molecules. It is highly expressed at endothelial cell junctions, and is also expressed in platelets and most leukocyte sub-types. The primary function of PECAM-1 is the mediation of leukocyte-endothelial cell adhesion and signal transduction. PECAM-1 has been implicated in the pathogenesis of various inflammation-related disorders, including thrombosis, multiple sclerosis (MS), and rheumatoid arthritis. The human PECAM-1 gene codes for a 738 amino acid transmembrane glycoprotein that contains a 118 amino acid cytoplasmic domain, a 19 amino acid transmembrane domain, and a 574 amino acid extracellular domain. Recombinant Human PECAM-1 is a 572 amino acid glycoprotein comprising the extracellular domain of PECAM-1. Monomeric glycosylated PECAM-1 migrates at an apparent molecular weight of approximately 80.0-95.0 kDa by SDS-PAGE analysis under reducing conditions. The calculated molecular weight of Recombinan Human PECAM-1 is 64.3 kDa.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty