Human Recombinant IL-24 (from CHO Cells)
Supplier: PEPROTECH INC.
200-35-5UG 200-35-20UG 200-35-50UG 200-35-100UG 200-35-250UG 200-35-500UG 200-35-1MG
10779-866EA 132.81 USD InStock
10779-866 10779-868 10779-870 10779-872 10779-874 10779-876 10779-878
Human Recombinant IL-24 (from CHO Cells)
Proteins and Peptides
IL-24 is a secreted glycoprotein belonging to the IL-10 structural family of cytokines. It is produced by a variety of cell types, including B cells, CD4+ cells, NK cells, lymph node DCs, monocytes, and melanoma cells. IL-24 can signal through the IL20R1/IL20R2 and IL22R1/IL20R2 receptors to initiate a signaling cascade, which includes the induction of JAK1/STAT3 phosphorylation. IL-24 is, functionally, a pleiotropic protein, but is generally characterized as an anti-cancer cytokine that can selectively inhibit growth of a wide variety of human cancer cells through activities that include: the induction of differentiation and apoptosis; and the suppression of angiogenesis and cell proliferation. Recombinant Human IL-24 is an 18.9 kDa glycoprotein that contains 161 amino acid residues, including a C-terminal His-tag.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty