Mouse Recombinant IL-31 (from E. coli)
Supplier: PEPROTECH INC.
210-31-2UG 210-31-10UG 210-31-100UG 210-31-250UG 210-31-500UG 210-31-1MG
10780-058EA 132.81 USD InStock
10780-058 10780-060 10780-062 10780-064 10780-066 10780-068
Mouse Recombinant IL-31 (from E. coli)
Proteins and Peptides
IL-31 is a T cell-derived cytokine that shares several structural and functional characteristics with IL-6, Oncostatin M, LIF, and Cardiotrophin-1. It signals through a receptor complex comprised of GPL (also known as gp130-like receptor or IL-31RA) and Oncostatin-M receptor (OSMR). GPL/OSMR signaling is a strong activator of STAT3 and STAT5, and can also activate STAT1, JAK1, and JAK2 signaling pathways. IL-31-regulated immune responses have been implicated in skin physiology and inflammatory skin diseases. Recombinant Murine IL-31 is a 15.8 kDa protein containing 141 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty