Human Recombinant HCC-1 (from E. coli)
Supplier: PEPROTECH INC.
300-38B-10UG 300-38B-100UG 300-38B-250UG 300-38B-500UG 300-38B-1MG
10780-528EA 338.16 USD InStock
10780-528 10780-530 10780-532 10780-534 10780-536
Human Recombinant HCC-1 (from E. coli)
Proteins and Peptides
HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. It is secreted by various tissues, including skeletal muscle, heart, spleen, liver, bone marrow and plasma. Mature HCC-1 is found in four different forms, which are distinguished by differential N-terminal truncation and contain 74, 72, 71, or 66 amino acid residues. Recombinant Human HCC-1 (CCL14) is a 7.8 kDa protein consisting of 66 amino acids, including the four highly conserved residues present in CC chemokines.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty