Human Recombinant Apo-SAA1 (from E. coli)
Supplier: PEPROTECH INC.
300-53-10UG 300-53-50UG 300-53-100UG 300-53-250UG 300-53-500UG 300-53-1MG
10780-572EA 132.81 USD InStock
10780-572 10780-574 10780-576 10780-578 10780-580 10780-582
Human Recombinant Apo-SAA1 (from E. coli)
Proteins and Peptides
Serum amyloid A proteins (SAA) represents a family of apolipoproteins that circulates in association with high-density lipoproteins (HDL). The level of Apo-SAA, normally 1-5 μg/ml in plasma, increases 500-1000 fold within 24 hours of an inflammatory stimulus and, under these conditions, is the most abundant HDL apolipoprotein. The human SAA gene codes for a 122 amino acid nonglycosylated polypeptide, which contains an 18 amino acid N-terminal sequence. Recombinant Human Apo-SAA1 is an 11.7 kDa protein containing 105 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty