Human Recombinant sCD100 (from CHO Cells)
Supplier: PEPROTECH INC.
310-29-250UG 310-29-500UG 310-29-1MG
10780-648EA 2806.2 USD InStock
10780-648 10780-650 10780-652
Human Recombinant sCD100 (from CHO Cells)
Proteins and Peptides
The Semphorins are a large family of phylogenetically conserved proteins that play a pivotal role in maintaining homeostasis in the immune system. Twenty members of this family have been identified and categorized into eight subclasses based on sequence similarity and distinctive structural features. CD100, also known as Sema4D, is a 150 kDa transmembrane class IV semaphorin. Studies have shown that CD100 can induce monocyte migration, T-cell activation, and B-cell survival, as well as T/B cell and T/DC “cooperation”. The CD100 precursor contains 862 amino acids, including a 21 a.a. signal sequence, a 713 a.a. extracellular domain, a 21 a.a. transmembrane sequence, and a 107 a.a. cytoplasmic region. The extracellular sequence contains several structural features, including a 479 a.a. “sema” domain, a 79 a.a. Ig-like sequence, and a 52 a.a. “Plexin-type repeat”. Recombinant Human sCD100 is a 78.9 kDa protein comprising the extracellular domain of CD100 (711 amino acids). SDS-PAGE analysis run under non-reducing conditions shows a mixture of disulfide linked dimer and monomer.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty