Mouse Recombinant Prolactin (from E. coli)
Supplier: PEPROTECH INC.
315-16-10UG 315-16-50UG 315-16-100UG 315-16-250UG 315-16-500UG 315-16-1MG
10780-744EA 132.81 USD InStock
10780-744 10780-746 10780-748 10780-750 10780-752 10780-754
Mouse Recombinant Prolactin (from E. coli)
Proteins and Peptides
Prolactin is a neuroendocrine hormone secreted by the pituitary gland. Its primary function is to promote and maintain lactation during pregnancy and suckling. In addition, prolactin plays an immunoregulatory role by stimulating the activities of ornithine decarboxylase and protein kinase C, which are important for the proliferation, differentiation, and function of lymphocytes. Recombinant Murine Prolactin is a 22.5 kDa globular protein containing 198 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty