Rat Recombinant IL-2 (from E. coli)
Supplier: PEPROTECH INC.
400-02-5UG 400-02-20UG 400-02-50UG 400-02-100UG 400-02-250UG 400-02-500UG 400-02-1MG
10780-870EA 132.81 USD InStock
10780-870 10780-872 10780-874 10780-876 10780-878 10780-880 10780-882
Rat Recombinant IL-2 (from E. coli)
Proteins and Peptides
IL-2 is a powerful immunoregulatory lymphokine produced by T-cells in response to antigenic or mitogenic stimulation. IL-2/IL-2R signaling is required for T-cell proliferation and other fundamental functions that are essential for the immune response. IL-2 stimulates growth and differentiation of B-cells, NK cells, lymphokine-activated killer cells, monocytes, macrophages and oligodendrocytes. Recombinant Rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues.
Información para pedidos: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty