Human Hepcidin-24 Trifluoroacetate
Supplier: Frontier Scientific
4084346.0500 4084346.1000
H-7378.0500BAEA 667.46 USD InStock
H-7378.0500BA H-7378.1000BA
Human Hepcidin-24 Trifluoroacetate
Proteins and Peptides
Hepcidin-24 is an excellent alternative to biotinylated or heavy isotope-labeled Hepcidin-25 used as internal reference in the determination of Hep-25 by mass spectrometry-based methods, e.g. in human serum or urine.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty