Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Formaldehyde 35.5 -38.0% stabilized ACS

Supplier: EPREDIA
Danger

Stabilization: Stabilized with methanol

15682 15680 15681
100503-402EA 49.55 USD InStock
100503-402 100503-908 100503-910
Formaldehyde 35.5 -38.0% stabilized ACS
Formaldehyde
HCHO F.W. 30.03 Specific Gravity: 1.09 A.C.S.

  • Specific Gravity: 1.09


Formula: CH₂O
Boiling Pt: 64 °C (147 °F)
Density: 1.09 g/ml
Flash Pt: 60 °C (140 °F
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 1198
ADR: 3,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d90419f0c71dbfa4a59794a6f4483bcedf092ec7&vid=d90419f0c71dbfa4a59794a6f4483bcedf092ec7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 35.5 to 38.0%
Residue after ignition 0.005%
Color (APHA) 10
Sulfate 0.002%
Heavy Metal (as Pb) 5ppm
Titratable acid 0.006 meq/g
Chloride (Cl) 5ppm
Iron (Fe) 5ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty