Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Formvar solution 1.00% in 1,2-dichloroethane

Supplier: EPREDIA

15830 15830-25
100503-940EA 40.72 USD InStock
100503-940 100502-402
Formvar solution 1.00% in 1,2-dichloroethane
Formvar
Formvar In Ethylene Dichloride 1% Solution
Microfiltered-ready to use.
Available in concentrations of 0.25%, 0.50% and 1.00%.


Formula: (C₃H₆O₂)n
MDL Number: MFCD00131103
CAS Number: 63148-64-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty