Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silver nitrate ≥99.9% ACS

Supplier: EPREDIA
Danger

21052 21050
100504-266EA 538.55 USD InStock
100504-266 100504-264
Silver nitrate ≥99.9% ACS
Silver nitrate
Reference: Elektronemikrokopische Swift, J.A. (1968). The electron histochemistry of cystine-containing proteins in thin transverse sections of human hair. J.R. Microsc. Soc. 88,449, Thiery, J.P. (1967). Mise en evidence des Polysaccharides sur coupes fines en microscopie eletronique. J. Microsc. (Paris) 6,987, Rambourg, A. (1967). An improved silver methenamine technique for the detection of periodic acid-reactive complex carbohydrates with the electron microscope.J. Histochem. Cytochem. 15, 409.

Hermandez et al., (1968)). Periodic acid-chromic acid-methenamine silver technique for glycoprotein detection in the electron microscope. J. Histochem. Cystochem. 16, 507., Ribi, W.A. (1976). A golgi-electron microscope method for insect nervous tissue. Stain Technol. 51,13., Braak, H., and Braak, E. (1982). A simple procedure for electron microscopy of Golgi-impregnated nerve cells. Neurosci. Lett. 32,1.


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 4.35 g/cm³ (36.301 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1 II
Merck Index: 12,08661
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4bc7a096e5fc163808f866e2392d908a14b7fe6&vid=d4bc7a096e5fc163808f866e2392d908a14b7fe6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99.9+%
Copper Max. 0.0002%
Iron Max. 0.0002%
Lead Max. 0.001%
Substances not precipitated by HCl Max. 0.010%
Sulfate Max. 0.0002%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty