Wilder Modification of Bielschowsky’s Method, Electron Microscopy Sciences
Supplier: EPREDIA
26392-02 26392-05 26392-06 26392-08 26392-10 26392-5A 26392-01 26392-07 26392-09 26392-03 26392-04
101411-328EA 31.55 USD InStock
101411-328 101411-334 101411-336 101411-340 101411-344 101411-346 101411-326 101411-338 101411-342 101411-330 101411-332
Wilder Modification of Bielschowsky’s Method, Electron Microscopy Sciences
Stains Histology Stains
Wilder Modification of Bielschowsky’s Method for staining of collagen and reticulum
Results - Reticulum Fibers (If beading observed and/or dark background, replace ammonium hydroxide silver, Nitrate solution & reducing solution): Black, Collagen: Rose.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty