Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LY-333,531 hydrochloride ≥98% (by HPLC)


270-348-M001 270-348-M005
89147-666EA 139 USD InStock
89147-666 89147-668
LY-333,531 hydrochloride ≥98% (by HPLC)
LY-333,531 hydrochloride
PKCβ inhibitor

Isozyme selective inhibitor of protein kinase Cβ (PKCβ). Inhibits both the PKCβI (IC50=4.7nM) and PKCβII (IC50=5.9nM) isozymes. 76- and 61-fold more selective for inhibition of PKCβI and PKCβII over PKCα.Not offered in the US.


Formula: C₂₈H₂₈N₄O₃·HCl
MDL Number: MFCD00940226
CAS Number: 169939-93-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty