Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dimethyl-2,2'-azodipropiononitrile ≥97%

Danger

086051-25G 086051-100G
102846-766EA 293.25 USD InStock
102846-766 102846-768
2,2'-Dimethyl-2,2'-azodipropiononitrile ≥97%
2,2'-Dimethyl-2,2'-azodipropiononitrile


Formula: C₈H₁₂N₄
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00013808
CAS Number: 78-67-1
UN: 3234
ADR: 4.1,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty