Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-SIGLEC7 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 194211]

Supplier: R&D Systems
FAB11381G-025 FAB11381G-100
102881-054EA 215.48 USD InStock
102881-054 102881-056
Anti-SIGLEC7 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 194211]
Antibodies
Type: Primary
Antigen: SIGLEC7
Clonality: Monoclonal
Clone: 194211
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty