Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CCR8 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 191704]

Supplier: R&D Systems
FAB1429A-025 FAB1429A-100
102881-206EA 244.4 USD InStock
102881-206 102881-208
Anti-CCR8 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 191704]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 191704
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty