Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-IL27RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 191106]

Supplier: R&D Systems
FAB14791A-025 FAB14791A-100
102880-482EA 316.71 USD InStock
102880-482 102880-484
Anti-IL27RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 191106]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 191106
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty