Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CXCR2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 242216]

Supplier: R&D Systems
FAB2164P-025 FAB2164P-100
102881-858EA 215.48 USD InStock
102881-858 102881-860
Anti-CXCR2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 242216]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 242216
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty