Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD4 Mouse Monoclonal Antibody (Fluorescein) [clone: 11830]

Supplier: R&D Systems
FAB3791F-025 FAB3791F-100
102882-578EA 99.78 USD InStock
102882-578 102882-580
Anti-CD4 Mouse Monoclonal Antibody (Fluorescein) [clone: 11830]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 11830
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty