Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FZD2 Rat Monoclonal Antibody [clone: 202102]

Supplier: R&D Systems
MAB1307-050 MAB1307-500
220018-983EA 360.09 USD InStock
220018-983 220018-984
Anti-FZD2 Rat Monoclonal Antibody [clone: 202102]
Antibodies
Type: Primary
Antigen Symbol: FZD2
Antigen Name: frizzled class receptor 2
Antigen Synonyms: Fz2, fz-2, fzE2, hFz2
Clonality: Monoclonal
Clone: 202102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty