Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-FZD7 Rat Monoclonal Antibody [clone: 151143]

Supplier: R&D Systems
MAB1981-100 MAB1981-050
220019-502EA 678.25 USD InStock
220019-502 220019-501
Anti-FZD7 Rat Monoclonal Antibody [clone: 151143]
Antibodies
Type: Primary
Antigen Symbol: FZD7
Antigen Name: frizzled homolog 7 (Drosophila)
Antigen Synonyms: Fz7
Clonality: Monoclonal
Clone: 151143
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty