Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Pyrimidin-2-yl)thiourea ≥97%

Supplier: Origen Biomedical, Inc
Warning

H33681-03 H33681-09
AAH33681-03EA 49.5 USD InStock
AAH33681-03 AAH33681-09
1-(Pyrimidin-2-yl)thiourea ≥97%
1-(Pyrimidin-2-yl)thiourea
Purity: 97+% Molecular Formula: C5H6N4S Formula Weight: 154.19 Melting Point: 234-238° MDL No.: MFCD02662218


Formula: C₅H₆N₄S
Storage Temperature: Ambient
MDL Number: MFCD02662218
CAS Number: 31437-20-4
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty