Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butoxy-6-chloropyridine 97%

Supplier: Origen Biomedical, Inc
Warning

H55085-03 H55085-06 H55085-14
AAH55085-03EA 23.5 USD InStock
AAH55085-03 AAH55085-06 AAH55085-14
2-tert-Butoxy-6-chloropyridine 97%
2-tert-Butoxy-6-chloropyridine
Purity: 97% CAS No.: 547740-43-2 Molecular Formula: C9H12ClNO Formula Weight: 185.65 Flash Point: 78°(172°F) Physical Form: Liquid Density: 1.076 Appearance: Colorless MDL No.: MFCD08705263


Formula: C₉H₁₂ClNO
Density: 1.076
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD08705263
CAS Number: 547740-43-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty