Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-methylbenzaldehyde 97%

Supplier: Origen Biomedical, Inc
Warning

H55228-03 H55228-06
AAH55228-03EA 124 USD InStock
AAH55228-03 AAH55228-06 AAH55228-MD
2-Fluoro-6-methylbenzaldehyde 97%
2-Fluoro-6-methylbenzaldehyde
Purity: 97% CAS No.: 117752-04-2 Molecular Formula: C8H7FO Formula Weight: 138.14 Physical Form: Liquid Density: 1.151 Refractive Index: 1.5250 Appearance: Colorless to yellow MDL No.: MFCD09258962


Formula: C₈H₇FO
Density: 1,151
Storage Temperature: Ambient
MDL Number: MFCD09258962
CAS Number: 117752-04-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty