Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-difluorobenzothiazole 97%

Supplier: Origen Biomedical, Inc
Warning

H55506-03 H55506-06
AAH55506-03EA 25.7 USD InStock
AAH55506-03 AAH55506-06 AAH55506-14
2-Amino-4,6-difluorobenzothiazole 97%
2-Amino-4,6-difluorobenzothiazole
Purity: 97% CAS No.: 119256-40-5 Molecular Formula: C7H4F2N2S Formula Weight: 186.18 Physical Form: Powder Appearance: White to pale brown Melting Point: 216-220° MDL No.: MFCD00276099


Formula: C₇H₄F₂N₂S
MW: 186.18
Melting Pt: 216…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00276099
CAS Number: 119256-40-5
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty