Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3:5,6-Di-O-isopropylidene-D-mannofuranose 98%

Supplier: Origen Biomedical, Inc

J66303-06 J66303-09 J66303-14
10815-506EA 62.1 USD InStock
10815-506 10815-508 10815-510
2,3:5,6-Di-O-isopropylidene-D-mannofuranose 98%
2,3:5,6-Di-O-isopropylidene-D-mannofuranose
Purity: 98% CAS No.: 14131-84-1 Molecular Formula: C12H20O6 Formula Weight: 260.28 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Melting Point: 125-126° dec. Beilstein Reference: 84382 MDL No.: MFCD00134206


Formula: C₁₂H₂₀O₆
Melting Pt: 125…126 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00134206
CAS Number: 14131-84-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty