Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Keto-D-gluconic acid hemicalcium salt monohydrate 99%, powder

Supplier: Origen Biomedical, Inc
Warning

J66609-03 J66609-06 J66609-09
10814-744EA 40.1 USD InStock
10814-744 10815-714 10815-716
2-Keto-D-gluconic acid hemicalcium salt monohydrate 99%, powder
2-Keto-D-gluconic acid hemicalcium salt monohydrate
Purity: 99% CAS No.: 3470-37-9 Molecular Formula: C6H9O7.1/2Ca.H2O Formula Weight: 231.22 Storage Temperature: 2°C to 8°C Physical Form: Powder Beilstein Reference: 3808734 Optical Rotation: -70° (c=2 in water) MDL No.: MFCD00149104


Formula: C6H9O7.1/2Ca .H2O
MW: 231.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00149104
CAS Number: 3470-37-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty