Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Butyldeoxynojirimycin ≥98%

Supplier: Origen Biomedical, Inc

J66613-LB0 J66613-MCR
10815-724EA 474 USD InStock
10815-724 10815-726
N-Butyldeoxynojirimycin ≥98%
N-Butyldeoxynojirimycin
An alpha-glucosidase inhibitor.

  • White Powder

Soluble in water at 10 mg/mL

An alpha-glucosidase Inhibitor


Formula: C₁₀H₂₁NO₄
Melting Pt: 126…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272581
CAS Number: 72599-27-0
Merck Index: 14,6188
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty