Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-β-D-xylopyranoside 98%

Supplier: Origen Biomedical, Inc

J66727-06 J66727-09 J66727-14
10814-802EA 76.44 USD InStock
10814-802 10814-804 10814-806
Methyl-β-D-xylopyranoside 98%
Methyl-beta-D-xylopyranoside
Purity: 98% CAS No.: 612-05-5 Molecular Formula: C6H12O5 Formula Weight: 164.16 Storage Temperature: -30°C to -10°C Physical Form: Powder Melting Point: 155-158° MDL No.: MFCD00047532


Formula: C₆H₁₂O₅
Melting Pt: 155…158 °C
Storage Temperature: Freezer
MDL Number: MFCD00047532
CAS Number: 612-05-5
EINECS: 210-289-7
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty