Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

beta-D-Thioglucose sodium salt 97%

Supplier: Origen Biomedical, Inc

J66838-03 J66838-MD
10815-888EA 156 USD InStock
10815-888 10815-890
beta-D-Thioglucose sodium salt 97%
beta-D-Thioglucose sodium salt
Purity: 97% CAS No.: 10593-29-0 Molecular Formula: C6H11NaO5S Formula Weight: 218.20 Storage Temperature: -30°C to -10°C Physical Form: Powder Solubility: Soluble in water MDL No.: MFCD00061630


Formula: C₆H₁₁NaO₅S
MW: 218.199 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00061630
CAS Number: 10593-29-0
EINECS: 234-200-6
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty