Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-O-Isopropylidene-D-erythronolactone 98%

Supplier: Origen Biomedical, Inc

J66871-03 J66871-06
10814-878EA 82.9 USD InStock
10814-878 10814-880
2,3-O-Isopropylidene-D-erythronolactone 98%
2,3-O-Isopropylidene-D-erythronolactone
For use in spiroannulated carbohydrate synthesis

Purity: 98% CAS No.: 25581-41-3 Molecular Formula: C7H10O4 Formula Weight: 158.15 Physical Form: Powder Appearance: White to off-white Melting Point: 67-69° Solubility: Soluble in water Beilstein Reference: 1282952 Optical Rotation: -118° (c=1 in water) MDL No.: MFCD00134440

For use in spiroannulated carbohydrate synthesis


Formula: C₇H₁₀O₄
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00134440
CAS Number: 25581-41-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty