Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anti-CD172a Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: P84]

Supplier: Biolegend
144005 144006
144005-BLEA 113.7 USD InStock
144005-BL 144006-BL
Anti-CD172a Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: P84]
Antibodies
FITC anti-mouse CD172a (SIRPα) [P84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD172a
Clonality: Monoclonal
Clone: P84
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty